Imprimer
Powered Vial Crimpers
Fournisseur: ADELPHI MANUFACTURING
6030503 6030504 6030520 6030505 6030514 6030507 6030508 6030509 6030500 6030501S 6030502 6030536 6030525/L
ADLM6030503EA 390.98 EUR
ADLM6030503 ADLM6030504 ADLM6030520 ADLM6030505 ADLM6030514 ADLM6030507 ADLM6030508 ADLM6030509 ADLM6030500 ADLM6030501S ADLM6030502 ADLM6030536 ADLM6030525/L
Powered Vial Crimpers
Pinces à sertir et pinces à dessertir
A fast and reliable air-operated power crimpers, for efficient crimping or decapping of vials.
  • Options for 'plain', 'flip top/flip off' and 'flip tear up' seals
  • Crimping and decapping of 13 and 20 mm seals
  • Fast and reliable for a professional quality finish
  • Simple and intuitive functionality
  • Corrosion-resistant construction
  • Air at 4 bar
The Powered Vial Crimper is available with stand and foot pedal and interchangeable screw-in heads. Foot pedal operation reduces operator fatigue. All crimping and de-capping heads are interchangeable with the Adelphi Manufacturing manual vial crimpers.
Operational weight: 9,85 kg
Shipping weight: 10,4 kg
Information de livraison: Supplied without head (available to purchase separately). Includes pressure regulator.