Imprimer
Pyridoxine hydrochloride ≥98% FCC
Fournisseur: Spectrum Chemical
Synonyms: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
P1459-100GM P1459-1KG
SPCMP1459-100GMEA 475 EUR
SPCMP1459-100GM SPCMP1459-1KG
Pyridoxine hydrochloride ≥98% FCC
Pyridoxine chlorhydrate
Pyridoxine Hydrochloride, FCC, a form of vitamin B6, is used as a vitamin B6 dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Point de fusion: 214…215 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00012807
Numéro CAS: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072
Commander maintenant
Résultats des tests de spécification
Assay 98% Min.