Imprimer
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% FCC
Fournisseur: Spectrum Chemical
BH104-125GM BH104-2.5KG BH104-45KGBL BH104-500GM
SPCMBH104-125GMEA 156 EUR
SPCMBH104-125GM SPCMBH104-2.5KG SPCMBH104-45KGBL SPCMBH104-500GM
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% FCC
tert-Butyl-4-méthoxyphénol (mélange d'isomères)
BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: C₁₁H₁₆O₂
Point d'ébullition: 264 °C (1013 hPa)
Point de fusion: 48…55 °C
Densité: 1,05 g/cm³ (20 °C)
Point d'éclair: 156 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD01779059
Numéro CAS: 25013-16-5
EINECS: 246-563-8
Merck Index: 12,01582
Commander maintenant
Résultats des tests de spécification
Assay 98.5% Min.