Imprimer
Titanium(IV) oxide ≥99% USP
Fournisseur: Spectrum Chemical
TI140-12KG TI140-500GM
SPCMTI140-12KGEA 837 EUR
SPCMTI140-12KG SPCMTI140-500GM
Titanium(IV) oxide ≥99% USP
Titane (IV) oxyde
Titanium Dioxide, USP is used as a food coloring. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: TiO₂
Point d'ébullition: 2900 °C (1013 hPa)
Point de fusion: 1855 °C
Densité: 4,2 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011269
Numéro CAS: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549
Commander maintenant
Résultats des tests de spécification
Assay 99% Min.