Imprimer
Cyclopentanone ≥99%
Fournisseur: Spectrum Chemical
C1124-200LTBL C1124-20LTBL
SPCMC1124-200LTBLEA 23830 EUR
SPCMC1124-200LTBL SPCMC1124-20LTBL
Cyclopentanone ≥99%
Cyclopentanone
Cyclopentanone is a liquid that has an odor similar to peppermint and is devoid of color. It's an organic compound and a cyclic ketone that most often used as a fragrance.
GHS hazard : Flame;Exclamation mark
UN2245
Formule: C₅H₈O
MW: 84.12 g/mol
Point d'ébullition: 130
Point de fusion: -58
Densité: 0,951
Point d'éclair: 31
Température de stockage: Température ambiante
Numéro MDL: MFCD00001409
Numéro CAS: 120-92-3
ONU: 2245
ADR: 3 III
Merck Index: 13,02771
Commander maintenant
Résultats des tests de spécification
Assay 99 % min.