Imprimer

Anti-Laminin 2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Fournisseur: Bioss
BOSSBS-8561R-A750EA 855 EUR
BOSSBS-8561R-A750
Anti-Laminin 2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Binding to cells via a high affinity receptor, laminin is thought to mediate the attachment, migration and organisation of cells into tissues during embryonic development by interacting with other extracellular matrix components.

Type: Primary
Antigen: Laminin 2 alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2051-2200/3122
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Bovine; Horse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR