Imprimer

Anti-Streptococcus suis Suilysin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Fournisseur: Bioss
BOSSBS-4537R-A680EA 855 EUR
BOSSBS-4537R-A680
Anti-Streptococcus suis Suilysin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Streptococcus suis suilysin belongs to a family of thiol-activated, membrane damaging toxins produced by some gram-positive bacteria. Although suilysin has been implicated as virulence associated, it has yet to be shown conclusively to be a virulence factor.

Type: Primary
Antigen: Streptococcus suis Suilysin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR