Imprimer
Septa for 20 mm stoppers, level 2 high-throughput applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
6A20ST1 6A20ST1F 6A20ST1T
HYPE6A20ST1EA 82.7 EUR
HYPE6A20ST1 HYPE6A20ST1F HYPE6A20ST1T
Septa for 20 mm stoppers, level 2 high-throughput applications, SureSTART™
Septa
Use Thermo Scientific™ SureSTART™ 20 mm stoppers with crimp vials that have a 20 mm opening and a secure push fit, including our SureSTART 20 mm crimp headspace vials (level 2).
Select level 2 products for high throughput applications, when robustness and reproducibility are key. Choose from various pack sizes of low permeability, 3 mm thick bromobutyl rubber stoppers designed to provide an effective alternative to crimp caps.