Imprimer
Brilliant III Ultra-Fast SYBR® green ROX QPCR master mix
Fournisseur: AGILENT
600889 600892 600903 600904
AGLS600889EA 361 EUR
AGLS600889 AGLS600892 AGLS600903 AGLS600904
Brilliant III Ultra-Fast SYBR® green ROX QPCR master mix
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
High performance, ultra-sensitive, SYBR® Green QPCR master mix reagent with ROX concentration for reliable quantification across a wide range of targets and templates.
  • Reliable and reproducible data
  • Novel fast Taq mutant for qPCR results in under 35 minutes Resistant to common QPCR inhibitors ( whole blood, humic acid, NaCl)
  • Enhanced rapid hot start capability saves time and reduces primer-dimer formation
  • Convenient pre-blended formulations compatible with any SYBR Green dye
Available with low or high ROX concentrations.
Commander maintenant
SPÉCIFICATIONS
Application qPCR dye
Environmentally Preferable