Imprimer
Pfu DNA polymerase
Fournisseur: AGILENT
600140 600153 600154 600159 600160
AGLS600140EA 1505 EUR
AGLS600140 AGLS600153 AGLS600154 AGLS600159 AGLS600160
Pfu DNA polymerase
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
High-quality polymerases for difficult to amplify DNA targets.
  • Proofreading DNA polymerase isolated from Pyrococcus furiosus
  • Ideal choice for many techniques requiring high-fidelity DNA synthesis by PCR
  • Blunt ends
Pfu DNA Polymerases includes Pfu DNA Polymerase, a proofreading DNA polymerase isolated from Pyrococcus furiosus.
The portfolio also features the recombinant Exo- Pfu DNA Polymerase (a genetically engineered mutant of the cloned Pfu DNA format) and the PfuTurbo DNA Polymerase with an optional alternative detergent.
Attention: For research use only. Not for use in diagnostic procedures.