Imprimer
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 97%
Fournisseur: Apollo Scientific
Attention
Synonyms: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA
PC0406-100G PC0406-25G PC0406-5G
APOSPC0406-100GEA 241 EUR
APOSPC0406-100G APOSPC0406-25G APOSPC0406-5G
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 97%
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
GC derivatising agent for silylation of hydroxy, carboxy, thio & primary and secondary amines.
Formule: C₉H₁₈F₃NOSi
Point d'ébullition: 168…170 °C
Densité: 1.023 g/cm³ (25 °C)
Point d'éclair: 52 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00009671
Numéro CAS: 77377-52-7
ONU: 1993
ADR: 3,III