Imprimer
IPTG (Isopropyl-β-D-thiogalactopyranoside) ≥99% sans dioxanne
Fournisseur: Apollo Scientific
Attention
Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
BIMB1008-25G BIMB1008-5G BIMB1008-10G
APOSBIMB1008-25GEA 87 EUR
APOSBIMB1008-25G APOSBIMB1008-5G APOSBIMB1008-10G
IPTG (Isopropyl-β-D-thiogalactopyranoside) ≥99% sans dioxanne
IPTG (Isopropyl-β-D-thiogalactopyranoside)
IPTG is used in conjunction with X-Gal for detection of lac+ colonies or cells in a colorimetric assay to detect recombinants (white) from non-recombinants (blue) in cloning strategies.
Formule: C₉H₁₈O₅S
MW: 238.31 g/mol
Point d'ébullition: 402
Point de fusion: 105 °C
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00063273
Numéro CAS: 367-93-1
EINECS: 206-703-0