Imprimer
β-D-Galactopyrannoside de 2-nitrophényle (ONPG) ≥99%
Fournisseur: Apollo Scientific
Attention
Synonyms: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
BIM1202-25G BIM1202-10G BIM1202-100G
APOSBIM1202-25GEA 71.2 EUR
APOSBIM1202-25G APOSBIM1202-10G APOSBIM1202-100G
β-D-Galactopyrannoside de 2-nitrophényle (ONPG) ≥99%
β-D-Galactopyrannoside de 2-nitrophényle (ONPG)
Colourimetric and spectrophotometric substrate for detection of b-Galactosidase activity. ONPG is cleaved byb-Galactosidase via hydrolysis at the b-1-4-glycosidicbond between 2-nitrophenol and galactose.
Formule: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Point de fusion: 192 °C (déc.)
Température de stockage: Congélateur
Numéro MDL: MFCD00063255
Numéro CAS: 369-07-3
EINECS: 206-716-1