Imprimer
Chlorophyllin, coppered trisodium salt, Sigma-Aldrich®
Fournisseur: SIGMA ALDRICH MICROSCOPY
C6003-100G C6003-25G C6003-5G
C6003-100GEA 380 EUR
C6003-100G C6003-25G C6003-5G
Chlorophyllin, coppered trisodium salt, Sigma-Aldrich®
Chlorophyllin, coppered trisodium salt
Chlorophyllin sodium copper salt has been used to study the effect of chlorophyllin against radiation-induced hematopoietic syndrome. It has been used to study the effect of chlorophyllin on the excretion of PhIP
Formule: C₃₄H₃₁CuN₄Na₃O₆
MW: 724.15 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00012149
Numéro CAS: 11006-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea436c5830499153985035352eee9262f0e7d056&vid=c824cb1f7d37e0910f9702dc6c491e3a920e0f13&prid=web&clid=SAPDAM