Imprimer
Disinfectant, Divosan Forte VT6
Fournisseur: Diversey
7510702 7510701
DIVY7510702EA 0 EUR
DIVY7510702 DIVY7510701
Disinfectant, Divosan Forte VT6
Produits nettoyants Désinfectants
Divosan Forte is a high-activity oxidising disinfectant based on peracetic acid for use in the food, beverage and dairy industries. Divosan Forte is a stabilised peracetic acid solution (15%) which is nonfoaming and completely free-rinsing.
  • Highly effective disinfectant against all types of microorganisms
  • Terminal disinfectant for use in automated CIP systems
  • Excellent deodorising and stain removal properties
  • Automatic injection using suitable CIP dosing equipment
  • Highly concentrated product for optimum cost-effectiveness
  • Suitable for use in soft or hard water
Versatile and effective CIP disinfectant, can be used in breweries, dairies, soft drinks plants and throughout the processed food industry. Powerful oxidising action also assists stain removal and deodorises. Free-rinsing and non-tainting ensures safe for all food applications.
Low environmental impact, breaks down to materials that are innocuous for waste water treatment.
Attention: Use biocides safely. Always read the label and product information before use.