Imprimer
Disinfectants, divodes FG, VT29
Fournisseur: Diversey
7516893 7514486
DIVY7516893EA 111 EUR
DIVY7516893 DIVY7514486
Disinfectants, divodes FG, VT29
Produits nettoyants Désinfectants
Divodes FG is a ready-to-use disinfectant spray for use in the food and beverage industry and also in the kitchen.
  • Ready to use (no dilution required)
  • Effective disinfection in a wide spectrum of applications
  • Non-foaming
  • Particularly recommended for use in areas sensitive to humidity
  • Evaporates quickly, no residue
Divodes FG is a fast acting disinfectant spray for intermediate and final treatment. It can be used safely on surfaces that come into contact with food and evaporate quickly without leaving visible residues. This spray allows the disinfection of alcohol-resistant surfaces, machines and components (conveyors, fillers, candles, water-sensitive components) as well as surfaces such as cutting tables generally encountered in the industry.
It is suitable for disinfecting displays in supermarkets, butcher shops, can be applied during intermediate breaks in order to maintain a stable hygiene standard until production restarts. It has a wide application spectrum and effectively removes bacteria and yeasts.
Attention: Use biocides safely. Always read the label and product information before use.