Imprimer

Xanthotoxin ≥98.0% (par GC)

Fournisseur: TCI
Danger

Synonyms: 9-Methoxy-7H-furo[3,2-g]chromen-7-one, 8-Methoxypsoralen, Methoxsalen

X0009-1G X0009-100MG
TCIAX0009-1GEA 46.2 EUR
TCIAX0009-1G TCIAX0009-100MG
Xanthotoxin ≥98.0% (par GC)
Xanthotoxin
Formule: C₁₂H₈O₄
Numéro MDL: MFCD00005009
Numéro CAS: 298-81-7
EINECS: 206-066-9
Merck Index: 14,05988

Commander maintenant

Résultats des tests de spécification

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C
Solubility in hot EtOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR