Imprimer

N-Vinyl-ε-caprolactam ≥98.0% (par GC) stabilisé

Fournisseur: TCI
Attention

Synonyms: 1-Vinylazepan-2-one

Stabilization: Stabilised with HO-TEMPO

V0073-100G V0073-500G
TCIAV0073-100GEA 39.6 EUR
TCIAV0073-100G TCIAV0073-500G
N-Vinyl-ε-caprolactam ≥98.0% (par GC) stabilisé
N-Vinyl-epsilon-caprolactam
Formule: C₈H₁₃NO
Numéro MDL: MFCD00080693
Numéro CAS: 2235-00-9
EINECS: 218-787-6

Commander maintenant

Résultats des tests de spécification

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR