Imprimer
Phycocyanin (de la spiruline)
Fournisseur: TCI
P2201-5G P2201-25G
TCIAP2201-5GEA 18.7 EUR
TCIAP2201-5G TCIAP2201-25G
Phycocyanin (de la spiruline)
Phycocyanin
Commander maintenant
Résultats des tests de spécification
Appearance Blue to Dark blue powder to crystal
Lambda max. 613.0 to 623.0 nm (buffer sol. pH6.0 Citric acid)
Absorbance (E10%1cm) min. 150 (buffer sol. pH6.0 Citric acid, 613.0 to 623.0 nm)
Suitability for protein-analysis to pass test