Imprimer
Aminomethyl polystyrene resin 2,0-3,0 mmol/g, "cross-linked" 1% DVB (200-400 mesh)
Fournisseur: TCI
A2047-25G A2047-5G
TCIAA2047-25GEA 314 EUR
TCIAA2047-25G TCIAA2047-5G
Aminomethyl polystyrene resin 2,0-3,0 mmol/g, "cross-linked" 1% DVB (200-400 mesh)
Aminomethyl polystyrene resin
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31b71be551d745ce9577f211e0ec90f522fc1a42&vid=b1cfe301f558e704143fbccbc7d9603b05359524&prid=web&clid=SAPDAM
Commander maintenant
Résultats des tests de spécification
Appearance White to Light yellow powder to crystalline
Loading 2.0 to 3.0 mmol/g