Imprimer
Acide hexachloroplatinique (IV) hexahydraté Pt ≥ 37,5% ≥99.95% (base métaux), Premion® faible teneur en nitrate
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate
42815.03 42815.06 42815.14
42815.03EA 170 EUR
42815.03 42815.06 42815.14
Acide hexachloroplatinique (IV) hexahydraté Pt ≥ 37,5% ≥99.95% (base métaux), Premion® faible teneur en nitrate
Acide hexachloroplatinique (IV) hexahydraté
Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.
Light sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.
Formule: H₂PtCl₆·6H₂O
MW: 517.9 g/mol
Point de fusion: 60 °C (déc.)
Densité: 2.430
Température de stockage: Température ambiante
Numéro MDL: MFCD00149910
Numéro CAS: 18497-13-7
EINECS: 241-010-7
ONU: 2507
ADR: 8 III
Merck Index: 14,7526