Imprimer
GC septa, BTO® septa
Fournisseur: Restek
27104 27117 27118 27115 27113 27112 27110 27100 27103 27101 27102 27105 27109 27106 27107 27108 27119
558-0409EA 0 EUR
558-0409 568-2385 568-2386 568-2383 568-2381 568-2380 568-2378 568-2369 568-2372 568-2370 568-2371 568-2373 568-2377 568-2374 568-2375 568-2376 568-2387
GC septa, BTO® septa
Septa
These septa are preconditioned and ready to use.
  • Usable to 400 °C inlet temperature
  • Precision moulding assures consistent, accurate fit
  • CenterGuide™ design requires less force for initial penetration
Bleed and temperature optimised, ideal for demanding GC and GC/MS applications.
Conditionnement: Packaged in non-contaminating glass jars.
Attention: Do not adhere to hot metal surfaces.