Imprimer
N,O-Bis(trimethylsilyl)acetamide: 1-(trimethylsilyl)imidazole: hexamethyldisilazane (BSA+TMSI+HMDS) mixture Silyl-271 réactif de silylation, pour dérivatisation GC
Fournisseur: MACHEREY-NAGEL
Danger
701450.110 701450.201 701450.510
MANA701450.110EA 64.7 EUR
MANA701450.110 MANA701450.201 MANA701450.510
N,O-Bis(trimethylsilyl)acetamide: 1-(trimethylsilyl)imidazole: hexamethyldisilazane (BSA+TMSI+HMDS) mixture Silyl-271 réactif de silylation, pour dérivatisation GC
N,O-Bis(trimethylsilyl)acetamide : 1-(trimethylsilyl)imidazole ; hexamethyldisilazane (BSA+TMSI+HMDS) mixture
Silyl-271 will derivatise all hydroxyl groups in any position. Useful in multiderivatisation schemes involving hydroxyl or amine groups.
  • Silylation can be applied on many compounds
  • Silylation reagents are easily prepared
  • Silyl-271 is well-suitable for compounds with different functional groups
BSA - HMDS - TSIM (2:7:1) silylation reagents without pyridine.
Point d'éclair: 10 °C
ONU: 3316
ADR: 9,II
IVDLogo