Imprimer
Fuchsine phéniquée solution, AFB-color colorant pour bactériologie, Sigma-Aldrich®
Fournisseur: Merck
Danger
Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride
M-Clarity™ (Niveau de Qualité) = MQ300
1.08512.0500 1.08512.2500
1.08512.0500EA 55.1 EUR
1.08512.0500 1.08512.2500
Fuchsine phéniquée solution, AFB-color colorant pour bactériologie, Sigma-Aldrich®
Fuchsine phéniquée
Tb-color carbol fuchsin solution is used to detect mycobacteria (AFB cold staining)
Tb-color carbol fuchsin solution is used for microscopic investigation of mycobacteria. This Tb-color staining kit is based on the classic Ziehl-Neelsen staining method. The Tb-color carbol fuchsin solution stains mycobacteria red in microscopic preparations, whereas other microorganisms take on the blue from the blue counter staining. The solution is an IVD product and CE registered.
Formule: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
Densité: 0.99 g/cm3 (20 °C)
Point d'éclair: 48 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00013129
Numéro CAS: 4197-24-4
ONU: 1992
ADR: 3 (6.1) III
IVDLogo