Imprimer
Silicon standard solution
Fournisseur: Bernd Kraft
03340.5020 03340.3000 03340.4000 03340.6020
KRAF03340.5020EA 162 EUR
KRAF03340.5020 KRAF03340.3000 KRAF03340.4000 KRAF03340.6020
Silicon standard solution
Étalons Étalons cationiques Standards de silicium
Sodium silicate in water can be used for determination of silicic acid in demineralised water.