Imprimer
ESD safety shoes, lace-up, DANIEL Pro Low
Fournisseur: ELTEN
7279101/46 7279101/47 7279101/48 7279102/40 7279102/41 7279102/42 7279102/43 7279102/44 7279102/45 7279102/46 7279102/47 7279102/48 7279103/40 7279103/41 7279103/42 7279103/43 7279103/44 7279103/45 7279103/46 7279103/47 7279103/48 7279101/40 7279101/41 7279101/42 7279101/43 7279101/44 7279101/45
ELTN7279101/46EA 91.9 EUR
ELTN7279101/46 ELTN7279101/47 ELTN7279101/48 ELTN7279102/40 ELTN7279102/41 ELTN7279102/42 ELTN7279102/43 ELTN7279102/44 ELTN7279102/45 ELTN7279102/46 ELTN7279102/47 ELTN7279102/48 ELTN7279103/40 ELTN7279103/41 ELTN7279103/42 ELTN7279103/43 ELTN7279103/44 ELTN7279103/45 ELTN7279103/46 ELTN7279103/47 ELTN7279103/48 ELTN7279101/40 ELTN7279101/41 ELTN7279101/42 ELTN7279101/43 ELTN7279101/44 ELTN7279101/45
ESD safety shoes, lace-up, DANIEL Pro Low
Chaussures et bottes
ESD safety shoes made from hydrophobised nubuck leather, fabric lining, reflective materials, a full padded bellows tongue and a dual density PU/PU sole ERGO-ACTIVE 2.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD black
  • Abrasion resistant scuff cap protects the leather upper
  • Anatomically-shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, form A