Imprimer
ESD safety shoes, lace-up, uvex 1 S2, 8534
Fournisseur: UVEX
8534.0/35 8534.0/41 8534.0/36 8534.0/37 8534.0/38 8534.0/39 8534.0/40 8534.0/42 8534.0/48 8534.0/44 8534.0/46 8534.0/49 8534.0/45 8534.0/43 8534.0/50 8534.0/51 8534.0/52 8534.0/47
UVEX8534.0/35EA 0 EUR
UVEX8534.0/35 UVEX8534.0/41 UVEX8534.0/36 UVEX8534.0/37 UVEX8534.0/38 UVEX8534.0/39 UVEX8534.0/40 UVEX8534.0/42 UVEX8534.0/48 UVEX8534.0/44 UVEX8534.0/46 UVEX8534.0/49 UVEX8534.0/45 UVEX8534.0/43 UVEX8534.0/50 UVEX8534.0/51 UVEX8534.0/52 UVEX8534.0/47
ESD safety shoes, lace-up, uvex 1 S2, 8534
Chaussures et bottes
Metal-free, extremely lightweight, flexible, sporty and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static). Free of lacquer-wetting inhibitors such as silicones or plasticisers.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome. Made from synthetic materials.
Colour: Black
Width: 14
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S2, SRC