Imprimer
VWR®, Multiwell Cell Culture Plates
Fournisseur: VWR Collection
10062-892 10062-894 10062-896
VWRU10062-892EA 0 EUR
VWRU10062-892 VWRU10062-894 VWRU10062-896
VWR®, Multiwell Cell Culture Plates
Microplaques Microplaques pour culture cellulaire
VWR® standard multiwell cell culture plates are plasma treated for consistent cell attachment and growth.
  • Clear polystyrene, sterile, certified non pyrogenic
  • Well volume uniformity ensures uniform growth surface area exposure
  • Single-position lid reduces misplacement and the risk of contamination
  • Designed with alphanumeric code for easy well identification
Raised rims on wells and condensation rings on the lid reduce evaporation and minimise edge effects.
Certifications: ISO 9001, ISO 13485.
Informations de commande: Individually wrapped in peel-to-open paper/plastic blister packs for all items.