Imprimer
Capillary columns, OPTIMA® 35 MS
Fournisseur: MACHEREY-NAGEL
726154.30 726154.60 726157.30 726157.60
MANA726154.30EA 680 EUR
MANA726154.30 MANA726154.60 MANA726157.30 MANA726157.60
Capillary columns, OPTIMA® 35 MS
Colonnes pour chromatographie
Medium polar silarylene GC phase with low bleeding.
  • Ideal for ion trap MS detectors
  • Recommended for environmental analyses, ultra-trace analyses, EPA methods, pesticides, PCBs, food and drug analyses, also as reference column in combination with 1 MS or 5 MS columns
  • Maximum temperature for isothermal operation 360 °C
  • Maximum temperature for short isotherms in a temperature program 370 °C
The selectivity is analogue to 35% phenyl - 65% methylpolysiloxane phases.
USP code G42 close equivalent to USP G28/G32.