Imprimer
ESD safety ankle boots, lace-up, Safety-Grip, Laurenzo Pu
Fournisseur: ELTEN
766841/43 766841/44 766841/45 766841/49 766841/50 766841/36 766841/37 766841/38 766841/39 766841/40 766841/41 766841/42 766841/46 766841/47 766841/48
ELTN766841/43EA 72.9 EUR
ELTN766841/43 ELTN766841/44 ELTN766841/45 ELTN766841/49 ELTN766841/50 ELTN766841/36 ELTN766841/37 ELTN766841/38 ELTN766841/39 ELTN766841/40 ELTN766841/41 ELTN766841/42 ELTN766841/46 ELTN766841/47 ELTN766841/48
ESD safety ankle boots, lace-up, Safety-Grip, Laurenzo Pu
Chaussures et bottes
Durable ESD safety ankle boots made from robust, breathable cowhide leather, a full padded bellows tongue plus a dual-density PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Full length fleece inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.