Imprimer
LEIC39602004
Fournisseur: Leica
Surgipath®
LEIC39602004EA 214 EUR
LEIC39602004
LEIC39602004
Paraplast Plus is suggested for large tissue or tissue that may normally present infiltration issues. Paraplast Plus contains a small percentage of dimethyl sulfoxide (DMSO), allowing accelerated tissue penetration. It has a melting point of 56° C.