Imprimer
N-Butanoyl-L-homoserine lactone 95, crystalline solid
Fournisseur: Cayman Chemical
Synonyms: N-Butyryl-L-Homoserine Lactone, C4-HSL, (S)-N-(2-Oxotetrahydrofuran-3-yl)butyramide, N-(2-Oxotetrahydro-3-furanyl)butanamide, N-Butanoyl-HoSer·Lactone
10007898-10 10007898-25 10007898-5 10007898-50
CAYM10007898-10EA 105 EUR
CAYM10007898-10 CAYM10007898-25 CAYM10007898-5 CAYM10007898-50
N-Butanoyl-L-homoserine lactone 95, crystalline solid
N-Butanoyl-L-homoserine lactone
Quorum sensing is a regulatory system used by bacteria for controlling gene expression in response to increasing cell density.
Controlling bacterial infections by quenching their quorum sensing systems is a promising field of study. The expression of specific target genes, such as transcriptional regulators belonging to the LuxIR family of proteins, is coordinated by synthesis of diffusible acylhomoserine lactone (AHL) molecules. N-butyryl-L-Homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and cellular metabolism.1 The diverse applications of this molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Formule: C₈H₁₃NO₃
MW: 171.2 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD12912262
Numéro CAS: 67605-85-0