Imprimer
Potassium hydrogénosulfate mixture of potassium pyrosulphate, K2S2O7, and potassium bisulphate, KHSO4 ACS, fondu
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Potassium bisulphate
13800.22 13800.36
13800.22EA 71.8 EUR
13800.22 13800.36
Potassium hydrogénosulfate mixture of potassium pyrosulphate, K2S2O7, and potassium bisulphate, KHSO4 ACS, fondu
Potassium hydrogénosulfate
Formula Weight: 136.17 Physical Form: Fused Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 214° UN No.: 2509 MDL No.: MFCD00066385
Formule: KHSO₄
MW: 136.17 g/mol
Point de fusion: 214
Densité: 2.32 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011404
Numéro CAS: 7646-93-7
EINECS: 231-594-1
ONU: 2509
ADR: 8 II
Merck Index: 13,07695