Imprimer
Tert-Butyle methyle oxyde, LiChrosolv® pour HPLC, Supelco®
Fournisseur: Merck
Danger
M-Clarity™ (Niveau de qualité) = MQ100
1.01845.1000 1.01845.2500 1.01845.9030 1.01845.9185
1.01845.1000EA 135 EUR
1.01845.1000 1.01845.2500 1.01845.9030 1.01845.9185
Tert-Butyle methyle oxyde, LiChrosolv® pour HPLC, Supelco®
Méthoxy-2-méthyl-2 propane
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.
Formule: (CH₃)₃COCH₃
Point d'ébullition: 55,2 °C (1013 hPa)
Point de fusion: –108,6 °C
Densité: 0,7405 g/cm³ (20 °C)
Point d'éclair: –28 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00008812
Numéro CAS: 1634-04-4
EINECS: 216-653-1
ONU: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM