Imprimer
Argent (I) acétate, anhydrous 99%
Fournisseur: Thermo Fisher Scientific
Attention
Synonyms: Silver acetate, Acetic acid silver (I) salt
11660.14 11660.22 11660.A1
11660.14EA 169 EUR
11660.14 11660.22 11660.A1
Argent (I) acétate, anhydrous 99%
Argent (I) acétate
Purity: 99% CAS No.: 563-63-3 Molecular Formula: C2H3AgO2 Formula Weight: 166.92 Physical Form: Crystalline Density: 3.26 Appearance: White to grey Odour: Odourless Melting Point: dec. Solubility: Soluble in dilute HNO3. Water solubility increases with temperature Merck Reference: 14,8505 UN No.: 3077 MDL No.: MFCD00012446
Formule: C₂H₃AgO₂
MW: 166.91 g/mol
Point de fusion: 2000 Déc.
Densité: 3.26
Température de stockage: Température ambiante
Numéro MDL: MFCD00012446
Numéro CAS: 563-63-3
EINECS: 209-254-9
ONU: 3077
ADR: 9 III
Merck Index: 14,8505