Imprimer
Ammonium formiate ≥98%
Fournisseur: Thermo Fisher Scientific
Attention
Synonyms: Formic acid ammonium salt
14517.18 14517.30
14517.18EA 65.9 EUR
14517.18 14517.30
Ammonium formiate ≥98%
Ammonium formiate
Purity: 98% CAS No.: 540-69-2 Molecular Formula: CH5NO2 Formula Weight: 63.06 Physical Form: Crystalline Density: 1.28 Appearance: White Odour: Ammonia-like Melting Point: ca 116° Beilstein Reference: 3625095 Merck Reference: 14,523 MDL No.: MFCD00013103
Formule: HCO₂NH₄
MW: 63.06 g/mol
Point de fusion: ∼116 °C
Densité: 1.28
Température de stockage: Température ambiante
Numéro MDL: MFCD00013103
Numéro CAS: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525