Imprimer
Chlorure de ruthenium (III) hydraté (≥38% Ru) ≥99.9% (base PGM)
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate
11043.18 11043.04 11043.09
11043.18EA 2850 EUR
11043.18 11043.04 11043.09
Chlorure de ruthenium (III) hydraté (≥38% Ru) ≥99.9% (base PGM)
Chlorure de ruthenium (III) hydraté
Purity: 99.9% CAS No.: 14898-67-0 Molecular Formula: Cl3Ru.xH2O Formula Weight: 207.43(anhy) Physical Form: Crystalline Soluble Sensitivity: Hygroscopic Appearance: Black Melting Point: 100°dec Solubility: Very soluble in water. Soluble in alcohol, acetone Merck Reference: 14,8302 UN No.: 3260 MDL No.: MFCD00149844
Oxidation
Formule: Cl₃Ru·xH₂O
MW: 207.43 g/mol (anhy)
Point de fusion: 100 °C (déc.)
Densité: 3.11 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149844
Numéro CAS: 14898-67-0
EINECS: 233-167-5
ONU: 3260
ADR: 8 III
Merck Index: 14,8302