Imprimer
Chrome (III) nitrate nonahydraté ≥98.5%
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate
11565.14 11565.30 11565.A1
11565.14EA 38.7 EUR
11565.14 11565.30 11565.A1
Chrome (III) nitrate nonahydraté ≥98.5%
Chrome (III) nitrate nonahydraté
Purity: 98.5% CAS No.: 7789-02-8 Molecular Formula: Cr(NO3)3.9H2O Formula Weight: 400.14 (238.01anhy) Physical Form: Crystalline Appearance: Purple Odour: Odourless Melting Point: 60° Solubility: Soluble in water, alcohol Merck Reference: 14,2226 UN No.: 2720 MDL No.: MFCD00149659
Catalyst, corrosion inhibitor, in textile printing
Formule: CrN₃O₉·9H₂O
MW: 400.15 g/mol
Point de fusion: 60
Densité: 1.807 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149659
Numéro CAS: 7789-02-8
EINECS: 236-921-1
ONU: 2720
ADR: 5.1 III
Merck Index: 13,02246