Imprimer

Sodium molybdate dihydraté

Fournisseur: Serva

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

30207.01 30207.02
SERA30207.01EA 0 EUR
SERA30207.01 SERA30207.02
Sodium molybdate dihydraté
Sodium molybdate dihydraté
Formule: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Point de fusion: 687
Densité: 3.28 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149170
Numéro CAS: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR