Imprimer
Ac-LEHD-AMC
Fournisseur: ENZO LIFE SCIENCES
ALX260080M001 ALX260080M005
ENZOALX260080M001EA 72.6 EUR
ENZOALX260080M001 ENZOALX260080M005
Ac-LEHD-AMC
Protéines et Peptides
Caspase-9 substrate
Fluorogenic substrate for caspase-9 and related cysteine proteases. The LEHD sequence is derived from combinatorial experiments. Ex.:340 to 360 nm, Em.:440 to 460 nm.