Imprimer
Anti-HSP60 Mouse Monoclonal Antibody
Fournisseur: ENZO LIFE SCIENCES
ADISPA806D ADISPA806F ADISPA806J
ENZOADISPA806DEA 290 EUR
ENZOADISPA806D ENZOADISPA806F ENZOADISPA806J
Anti-HSP60 Mouse Monoclonal Antibody
Anticorps
Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.
  • For Immunofluorescence, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus HSP60. Detects a band of ~60kDa by Western blot.
Type: Primary
Antigen: HSP60
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human