Imprimer
Capillary columns, Pora-PLOT Q
Fournisseur: Agilent
CP7551PT CP7550PT
HEWLCP7551PTEA 906 EUR
HEWLCP7551PT HEWLCP7550PT
Capillary columns, Pora-PLOT Q
Colonnes pour chromatographie
These Pora-PLOT Q columns are mostly recommended for the column switching systems that involves in the analysis of polar and apolar volatile compounds.
  • Available in fused silica and ultiMetal
  • Long term stability provides repeatable retention times
  • Water elutes as a sharp peak enabling quantitation
Each of these type of columns are tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results.
Informations de commande: Similar phases are available as Rt-Q BOND, Rt-QPLOT, SupelQ PLOT columns.