Imprimer
Peel-A-Way® Embedding Moulds
Fournisseur: POLYSCIENCES
18646A-1 18646B-1 18646C-1 18986-1 18646ACODE45 18986CODE42 18646D-1
POLS18646A-1EA 0 EUR
POLS18646A-1 POLS18646B-1 POLS18646C-1 POLS18986-1 POLS18646ACODE45 POLS18986CODE42 POLS18646D-1
Peel-A-Way® Embedding Moulds
Moules Moules pour l'histologie
These disposable paraffin moulds are designed to 'Peel-A-Way' from the solidified embedded block.
  • Versatile and can be used with polyfreeze, JB-4, GMA, MMA and epoxy
  • Allows the specimen to freeze to the chuck without defrosting the base
  • Unique polyethylene construction eliminates the need for release agents and block trimming
Useful for frozen tissue where defrosting during transfer to cryostat is an issue.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable