Imprimer
Tubing, PVC
Fournisseur: VEMOPLEX
VP1013.0010 VP1013.0050 VP1014.0010 VP1014.0050 VP1016.0050
228-0891EA 110 EUR
228-0891 228-0892 228-0893 228-0894 VEMOVP1016.0050
Tubing, PVC
Tuyaux Transfer Tubing
PVC tubing is used for transporting liquids and gases in installations without pressure, where it is important that the fluid be visible. Suitable for food products following BGA.
  • Construction: One-layer soft PVC hose
  • Smooth inner and outer wall
  • Colourless, transparent
Temperature range: –15 to 50 °C.