Imprimer

Anti-EAST1 Rabbit Polyclonal Antibody (Cy5®)

Fournisseur: Bioss
BOSSBS-8858R-CY5EA 858 EUR
BOSSBS-8858R-CY5
Anti-EAST1 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
heatstable enterotoxin 1 [Escherichia coli]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EAST1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR