Imprimer

Anti-streptococcus suis Suilysin Rabbit Polyclonal Antibody (Cy3®)

Fournisseur: Bioss
BOSSBS-4537R-CY3EA 858 EUR
BOSSBS-4537R-CY3
Anti-streptococcus suis Suilysin Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Streptococcus suis serotype 2 (SS2) infection has become a public health concern due to its zoonotic capability to cause severe infections in slaughterhouse workers and those who handle infected pork.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: streptococcus suis Suilysin
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR