Imprimer
Diméthylsulfoxyde 99 Cell Culture
Fournisseur: MP Biomedicals
Attention
Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
0219605525 0219605580 0219605590 0219605591
ICNA0219605525EA 37.5 EUR
ICNA0219605525 ICNA0219605580 ICNA0219605590 ICNA0219605591
Diméthylsulfoxyde 99 Cell Culture
Diméthylsulfoxyde
Dimethyl Sulfoxide is ideal for use as a cryoprotectant. Also used in chemical reactions, in polymerase chain reactions (PCR) as a PCR cosolvent to help improve yields, especially in long PCR, and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal-induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells (ESC), and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum (FBS). Used to enhance dermal absorption of many chemicals. A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers. Used in antifreeze or hydraulic fluids. A cryopreservative for cell cultures. Used in the oxidation of thiols and disulfides to sulfonic acids.
Dimethyl sulfoxide (DMSO) is a highly polar aprotic organic reagent that has exceptional solvent properties for organic and inorganic chemicals.
Physical Apppearance: Liquid
Viscosity: 1.1 centipoises (27 °C)
Vapor Density: 2.7 (vs air)
Vapor Pressure: 0.55 hPa at 20 °C, 0.42 mmHg (20 °C)
Refractive Index: n20/D 1.479(lit.)
Solubility: Soluble in water (1 mL DMSO + 1 mL H2O), methanol, acetone, ether, benzene, chloroform
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Point d'ébullition: 189 °C (lit.)                      
Point de fusion: 16…19 °C (lit.)                      
Densité: 1.104 g/cm³ (20 °C)
Point d'éclair: 87 °C (188.6 °F) (coupe fermée)
Température de stockage: Température ambiante
Numéro MDL: MFCD00002089
Numéro CAS: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM