Imprimer
Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol) ≥99%
Fournisseur: MP Biomedicals
Attention
02152176.1 02152176.5 0215217601 0215217605 0215217610
ICNA02152176.1EA 36.1 EUR
ICNA02152176.1 ICNA02152176.5 ICNA0215217601 ICNA0215217605 ICNA0215217610
Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol) ≥99%
Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol)
TRIS (Trometamol, Tromethamine, Tris-[hydroxymethyl]aminomethane) Purity: 99.0-99.5% Buffering pH range 7.0-9.0 pKa at 25°C = 8.1 This grade of tris is excellent where purity and value are both important. It is superior to our technical grade tris and less expensive than our ultra pure material. This grade will suffice for most buffer applications.
pH: 10 - 11.5 (5% aq soln)
Formule: NH₂C(CH₂OH)₃
Température de stockage: Température ambiante
Numéro MDL: MFCD00004679
Numéro CAS: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM