Imprimer
Flame retardant overalls, MICROGARD® 1500 PLUS FR, model 111
Fournisseur: Ansell
WR17S-00111-03 WR17S-00111-04 WR17S-00111-05 WR17S-00111-06 WR17S-00111-07
ANSEWR17S-00111-03EA 333 EUR
ANSEWR17S-00111-03 ANSEWR17S-00111-04 ANSEWR17S-00111-05 ANSEWR17S-00111-06 ANSEWR17S-00111-07
Flame retardant overalls, MICROGARD® 1500 PLUS FR, model 111
Vêtements à usage unique
These overalls are made from a highly breathable, flame retardant and anti-static SMMS polypropylene non woven fabric designed for protection from particulates and light, non flammable liquid spray or splash.
  • Three-piece hood
  • Two-way front zipper with resealable storm flap; finger loops
  • Elasticated wrists, waist and ankles
  • Red stitching for ease of identification
  • Stitched seams combine strength with particle barrier
  • Flame retardant and anti-static SMMS provides a good barrier to particulates and low hazard liquid sprays or splashes
  • Air and water vapour permeable ('breathable') to help reduce the risk of heat stress
Intended to be worn over heat and flame protective clothing, MICROGARD® 1500 PLUS FR offers wearers protection from particulates (Type 5) and non flammable, non pressurised liquid sprays or splashes (Type 6).*
Typical applications include: Petrochemical industry, industrial cleaning, utilities, general maintenance.
Certifications: Type 5/6; EN 1073-2; EN ISO 14116 (index 1/0/0), EN 1149-5; Cat. III
Attention: Must be worn over thermal protective garments (which achieve EN ISO 14116 Index 2 or above), such as NOMEX® or PYROVATEX®, and never be worn next to the skin.