Imprimer
Guard/retention gap columns, hydroguard® treated MXT® (passivated stainless steel)
Fournisseur: Restek
70080 70081 70083 70084 70086 70087
REST70080EA 0 EUR
REST70080 REST70081 REST70083 REST70084 REST70086 REST70087
Guard/retention gap columns, hydroguard® treated MXT® (passivated stainless steel)
Colonnes pour chromatographie
Extend analytical column lifetime by preventing degradation from harsh 'steam-cleaning' water injections.
Maximum temperature: 325 °C